Merck
  • A laboratory practical illustrating the use of the ChIP-qPCR method in a robust model: Estrogen receptor alpha immunoprecipitation using Mcf-7 culture cells.
A laboratory practical illustrating the use of the ChIP-qPCR method in a robust model: Estrogen receptor alpha immunoprecipitation using Mcf-7 culture cells.
Biochemistry and molecular biology education : a bimonthly publication of the International Union of Biochemistry and Molecular Biology (2016-09-27)
Eric Lacazette
ABSTRACT
Chromatin immunoprecipitation followed by qPCR analysis (ChIP-qPCR) is a widely used technique to study gene expression. A large number of students in molecular biology and more generally in life sciences will be confronted with the use of this technique, which is quite difficult to set up and can lead to misinterpretation if not carefully managed. This article describes a four-session laboratory practical designed for Masters students to introduce this technique. During the practical, students work in pairs. They extract chromatin from MCF-7 culture cells stimulated or not by estrogens, then immunoprecipitate the transcription factor estrogen receptor alpha using an antibody directed against it. Students then measure the enrichment of promoter DNA target sequences from the chromatin that coprecipitates by qPCR. These are two estrogen responsive genes, pS2 (trefoil factor one) and PGR (the progesterone receptor). They learn how to analyze their ChIP-qPCR data by two methods: percent input and fold enrichment, and are taught about the interpretation limits of these two calculation methods. Thus, this practical is a good framework for an in-depth discussion of how this technique can be used to study gene expression and for raising awareness of the importance of careful interpretation of results. © 2016 by The International Union of Biochemistry and Molecular Biology, 45(2):152-160, 2017.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Lithium chloride, for molecular biology, ≥99%
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With 15 mM HEPES and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Deoxyribonucleic acid sodium salt from salmon testes
Sigma-Aldrich
Monoclonal Anti-HA antibody produced in mouse, clone HA-7, ascites fluid
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks