Merck
  • Quantitative structure-activity relationship and complex network approach to monoamine oxidase A and B inhibitors.
Quantitative structure-activity relationship and complex network approach to monoamine oxidase A and B inhibitors.
Journal of medicinal chemistry (2008-10-07)
Lourdes Santana, Humberto González-Díaz, Elías Quezada, Eugenio Uriarte, Matilde Yáñez, Dolores Viña, Francisco Orallo
ABSTRACT
The work provides a new model for the prediction of the MAO-A and -B inhibitor activity by the use of combined complex networks and QSAR methodologies. On the basis of the obtained model, we prepared and assayed 33 coumarin derivatives, and the theoretical prediction was compared with the experimental activity data. The model correctly predicted 27 compounds, and most of the active derivatives showed IC 50 values in the muM-nM range against both the MAO-A and MAO-B isoforms. Compound 14 shows the same MAO-A inhibitory activity (IC 50 = 7.2 nM), as clorgyline used as a reference inhibitor and has the highest MAO-A specificity (1000-fold higher compared to MAO-B). On the other hand, compounds 24 (IC 50 = 1.2 nM) and 28 (IC 50 = 1.5 nM) show higher activity than selegiline (IC 50 = 19.6 nM) and high MAO-B selectivity with 100-fold and 1600-fold inhibition levels, with respect to the MAO-A isoform.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Razoxane, >98% (HPLC)
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Cycloheximide, PESTANAL®, analytical standard
Supelco
Ibuprofen
Supelco
Lindane, PESTANAL®, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
MCPA, PESTANAL®, analytical standard
Supelco
Sulfamethoxypyridazine, VETRANAL®, analytical standard
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Sigma-Aldrich
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
Supelco
Linuron, PESTANAL®, analytical standard
Sigma-Aldrich
Resorcinol, ≥98%, FG
Sigma-Aldrich
2,2′-Thiodiethanol, ≥99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Resorcinol, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
Sigma-Aldrich
3-Methyl-1-pentyn-3-ol, 98%