Merck
  • Novel potent and selective bile acid derivatives as TGR5 agonists: biological screening, structure-activity relationships, and molecular modeling studies.
Novel potent and selective bile acid derivatives as TGR5 agonists: biological screening, structure-activity relationships, and molecular modeling studies.
Journal of medicinal chemistry (2008-03-01)
Hiroyuki Sato, Antonio Macchiarulo, Charles Thomas, Antimo Gioiello, Mizuho Une, Alan F Hofmann, Régis Saladin, Kristina Schoonjans, Roberto Pellicciari, Johan Auwerx
ABSTRACT
TGR5, a metabotropic receptor that is G-protein-coupled to the induction of adenylate cyclase, has been recognized as the molecular link connecting bile acids to the control of energy and glucose homeostasis. With the aim of disclosing novel selective modulators of this receptor and at the same time clarifying the molecular basis of TGR5 activation, we report herein the biological screening of a collection of natural occurring bile acids, bile acid derivatives, and some steroid hormones, which has resulted in the discovery of new potent and selective TGR5 ligands. Biological results of the tested collection of compounds were used to extend the structure-activity relationships of TGR5 agonists and to develop a binary classification model of TGR5 activity. This model in particular could unveil some hidden properties shared by the molecular shape of bile acids and steroid hormones that are relevant to TGR5 activation and may hence be used to address the design of novel selective and potent TGR5 agonists.
Product Description
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Glycoursodeoxycholic acid, ≥96.0% (TLC)
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard
Sigma-Aldrich
5α-Pregnane-3,20-dione (allo), progesterone metabolite
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
trans-Androsterone
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Supelco
Androsterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Hyodeoxycholic acid, ≥98%
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Etiocholan-3α-ol-17-one
Sigma-Aldrich
5α-Androstane-3α,17β-diol
Supelco
Testosterone, VETRANAL®, analytical standard
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%
Sigma-Aldrich
Ursodeoxycholic acid, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture