Merck
A mosaic analysis system with Cre or Tomato expression in the mouse.
Proceedings of the National Academy of Sciences of the United States of America (2020-10-28)
Qun Wang, Yen-Yu Lin, Baojun Zhang, Jianxuan Wu, Sumedha Roy, Jeremy J Ratiu, Yanping Xu, Meifang Dai, Laura P Hale, Yue Xiong, Qi-Jing Li, Yuan Zhuang
ABSTRACT
Somatic mutations are major genetic contributors to cancers and many other age-related diseases. Many disease-causing somatic mutations can initiate clonal growth prior to the appearance of any disease symptoms, yet experimental models that can be used to examine clonal abnormalities are limited. We describe a mosaic analysis system with Cre or Tomato (MASCOT) for tracking mutant cells and demonstrate its utility for modeling clonal hematopoiesis. MASCOT can be induced to constitutively express either Cre-GFP or Tomato for lineage tracing of a mutant and a reference group of cells simultaneously. We conducted mosaic analysis to assess functions of the Id3 and/or Tet2 gene in hematopoietic cell development and clonal hematopoiesis. Using Tomato-positive cells as a reference population, we demonstrated the high sensitivity of this system for detecting cell-intrinsic phenotypes during short-term or long-term tracking of hematopoietic cells. Long-term tracking of Tet2 mutant or Tet2/Id3 double-mutant cells in our MASCOT model revealed a dynamic shift from myeloid expansion to lymphoid expansion and subsequent development of lymphoma. This work demonstrates the utility of the MASCOT method in mosaic analysis of single or combined mutations, making the system suitable for modeling somatic mutations identified in humans.
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, lyophilized powder, Protein ≥85 %, ≥400 Kunitz units/mg protein
Pricing and availability is not currently available.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Pricing and availability is not currently available.
Sigma-Aldrich
Percoll®, Cytiva 17-0891-01, pack of 1 L
Pricing and availability is not currently available.