Merck
In vitro anticoccidial activity of thymol, carvacrol, and saponins.
Poultry science (2020-11-05)
Martina Felici, Benedetta Tugnoli, Federico Ghiselli, Paola Massi, Giovanni Tosi, Laura Fiorentini, Andrea Piva, Ester Grilli
ABSTRACT
The anticoccidial activity of thymol, carvacrol, and saponins was assessed in an in vitro model of coccidiosis. Eimeria spp. sporozoites were collected from field samples, characterized, and used for 2 different invasion assays on Madin-Darby Bovine Kidney cells (MDBK). The cells were challenged with 5 × 104 sporozoites without (control) or with various treatments: saponins (10 ppm), thymol, and carvacrol (7 ppm each) or a combination of saponins, thymol, and carvacrol at 2 doses; MIX 1 (saponins 5 ppm, thymol 3.5 ppm, and carvacrol 3.5 ppm) and MIX 2 (saponins 10 ppm, thymol 7 ppm, and carvacrol 7 ppm). The treated cells were incubated at 37°C for 24 h (invasion assay 1) and for 2, 24, and 48 h (invasion assay 2). The efficiency of invasion was determined by counting the sporozoites left in the supernatant that were not able to invade the cells, whereas intracellular Eimeria DNA was detected by qPCR to confirm the data. Data were analyzed with ANOVA, and differences were considered significant when P value was ≤0.05. Data from invasion assay 1 showed that the thymol and carvacrol-containing blends significantly reduced invasion, especially in combination with saponins at the highest dose. Saponins alone did not have a strong inhibiting activity but acted synergistically with the other molecules. Interestingly, in invasion assay 2, it was found that the effect of the highest dose of the blend of saponins, thymol, and carvacrol was already visible at 2 h postinfection, whereas the other treatments were significantly successful at 24 h postinfection. The invasion assay protocol was designed to screen molecules in vitro starting from field fecal samples, and it can represent a potential tool in Eimeria research. Moreover, this study shows that invasion in MDBK cells by Eimeria sporozoites is inhibited in presence of thymol, carvacrol, and saponins, thus highlighting the anticoccidial potential of these compounds.
Product Description
Millipore
Bile salts, suitable for microbiology, (extracted under controlled conditions from purified fresh bile)
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose and sodium bicarbonate, without L-glutamine, sodium pyruvate, and phenol red, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Carvacrol, natural, 99%, FG
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Thymol, ≥98.5%
Sigma-Aldrich
Potassium dichromate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture