Merck
  • Evidence for a Solid-Electrolyte Inductive Effect in the Superionic Conductor Li10Ge1-xSnxP2S12.
Evidence for a Solid-Electrolyte Inductive Effect in the Superionic Conductor Li10Ge1-xSnxP2S12.
Journal of the American Chemical Society (2020-12-08)
Sean P Culver, Alexander G Squires, Nicolò Minafra, Callum W F Armstrong, Thorben Krauskopf, Felix Böcher, Cheng Li, Benjamin J Morgan, Wolfgang G Zeier
ABSTRACT
Strategies to enhance ionic conductivities in solid electrolytes typically focus on the effects of modifying their crystal structures or of tuning mobile-ion stoichiometries. A less-explored approach is to modulate the chemical bonding interactions within a material to promote fast lithium-ion diffusion. Recently, the idea of a solid-electrolyte inductive effect has been proposed, whereby changes in bonding within the solid-electrolyte host framework modify the potential energy landscape for the mobile ions, resulting in an enhanced ionic conductivity. Direct evidence for a solid-electrolyte inductive effect, however, is lacking-in part because of the challenge of quantifying changes in local bonding interactions within a solid-electrolyte host framework. Here, we consider the evidence for a solid-electrolyte inductive effect in the archetypal superionic lithium-ion conductor Li10Ge1-xSnxP2S12. Substituting Ge for Sn weakens the {Ge,Sn}-S bonding interactions and increases the charge density associated with the S2- ions. This charge redistribution modifies the Li+ substructure causing Li+ ions to bind more strongly to the host framework S2- anions, which in turn modulates the Li+ ion potential energy surface, increasing local barriers for Li+ ion diffusion. Each of these effects is consistent with the predictions of the solid-electrolyte inductive effect model. Density functional theory calculations predict that this inductive effect occurs even in the absence of changes to the host framework geometry due to Ge → Sn substitution. These results provide direct evidence in support of a measurable solid-electrolyte inductive effect and demonstrate its application as a practical strategy for tuning ionic conductivities in superionic lithium-ion conductors.
Sigma-Aldrich
Lithium sulfide, 99.98% trace metals basis
Sigma-Aldrich
Germanium(II) sulfide, 99.99% trace metals basis