Merck
  • Quantitative structure-activity relationship studies for prediction of antimicrobial activity of synthesized 2,4-hexadienoic acid derivatives.
Quantitative structure-activity relationship studies for prediction of antimicrobial activity of synthesized 2,4-hexadienoic acid derivatives.
Bioorganic & medicinal chemistry letters (2007-09-18)
Balasubramanian Narasimhan, Vikramjeet Judge, Rakesh Narang, Ruchita Ohlan, Sucheta Ohlan
ABSTRACT
A new series of 2,4-hexadienoic acid derivatives (S(1)-S(42)) has been synthesized and evaluated as antimicrobial agents against Staphylococcus aureus, Bacillus subtilis, Escherichia coli, Candida albicans, and Aspergillus niger. Quantitative structure-activity relationship (QSAR) investigation using Hansch analysis was applied to find out correlation between antimicrobial activities with physicochemical properties of the synthesized compounds. Various physicochemical descriptors and experimentally determined minimum inhibitory concentration values for different microorganisms were used as independent and dependent variables, respectively. The QSAR revealed that topological parameters especially molecular connectivity indices (chi(2), (0)chi(v), (2)chi(v)) were found to have overall significant correlation with antimicrobial activity of 2,4-hexadienoic acid derivatives. The statistical results of training set, cross-validated r(2) and conventional r values gave reliability to the prediction of molecules with activity using QSAR models.
Product Description
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sigma-Aldrich
Ethyl sorbate, ≥97%, FG