Merck
  • Subcellular NAMPT-mediated NAD+ salvage pathways and their roles in bioenergetics and neuronal protection after ischemic injury.
Subcellular NAMPT-mediated NAD+ salvage pathways and their roles in bioenergetics and neuronal protection after ischemic injury.
Journal of neurochemistry (2019-09-26)
Xiaowan Wang, Zhe Zhang, Nannan Zhang, Hailong Li, Li Zhang, Christopher P Baines, Shinghua Ding
ABSTRACT
NAD+ is a cofactor required for glycolysis, tricarboxylic acid cycle, and complex I enzymatic reaction. In mammalian cells, NAD+ is predominantly synthesized through the salvage pathway, where nicotinamide phosphoribosyltransferase (NAMPT) is the rate-limiting enzyme. Previously, we demonstrated that NAMPT exerts a neuroprotective effect in ischemia through the suppression of mitochondrial dysfunction. Mammalian cells maintain distinct NAD+ pools in the cytosol, mitochondria, and nuclei. However, it is unknown whether mitochondria have an intact machinery for NAD+ salvage, and if so, whether it plays a dominant role in bioenergetics, mitochondrial function, and neuronal protection after ischemia. Here, using mouse primary cortical neuron and cortical tissue preparations, and multiple technologies including cytosolic and mitochondrial subfractionation, viral over-expression of transgenes, molecular biology, and confocal microscopy, we provided compelling evidence that neuronal mitochondria possess an intact machinery of NAMPT-mediated NAD+ salvage pathway, and that NAMPT and nicotinamide mononucleotide adenylyltransferase 3 (NMNAT3) are localized in the mitochondrial matrix. By knocking down NMNAT1-3 and NAMPT with siRNA, we found that NMNAT3 has a larger effect on basal and ATP production-related mitochondrial respiration than NMNAT1-2 in primary cultured neurons, while NMNAT1-2 have a larger effect on glycolytic flux than NMNAT3. Using an oxygen glucose deprivation model, we found that mitochondrial, cytoplasmic, and non-subcellular compartmental over-expressions of NAMPT have a comparable effect on neuronal protection and suppression of apoptosis-inducing factor translocation. The current study provides novel insights into the roles of subcellular compartmental NAD+ salvage pathways in NAD+ homeostasis, bioenergetics, and neuronal protection in ischemic conditions.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Anti-Actin antibody, Mouse monoclonal, clone AC-40, purified from hybridoma cell culture
Sigma-Aldrich
Anti-AIF antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-C1QBP antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection
Sigma-Aldrich
Donkey Anti-Rabbit IgG Antibody, Rhodamine conjugate, Species Adsorbed, Chemicon®, from donkey
Sigma-Aldrich
Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, The Nicotinamide Phosphoribosyltransferase Inhibitor, FK866, also referenced under CAS 658084-64-1, controls the biological activity of Nicotinamide Phosphoribosyltransferase. This small molecule/inhibitor is primarily used for Neuroscience applications.