Merck
  • Synthesis and antimicrobial evaluation of new chalcones containing piperazine or 2,5-dichlorothiophene moiety.
Synthesis and antimicrobial evaluation of new chalcones containing piperazine or 2,5-dichlorothiophene moiety.
Bioorganic & medicinal chemistry letters (2007-08-28)
V Tomar, G Bhattacharjee, Kamaluddin, Ashok Kumar
ABSTRACT
Two new series of chalcones have been synthesized by reacting 1-(4-piperazin-1-yl-phenyl)ethanone and 1-(2,5-dichloro-3-thienyl)-1-ethanone with different substituted benzaldehydes in turn by Claisen-Schmidt condensation. The compounds have been characterized by IR, (1)H NMR spectral and microanalysis data. All the synthesized compounds have been evaluated for antimicrobial activity. Some of these derivatives are potentially active against Gram-positive bacteria, Staphylococcus aureus and Escherichia coli while the most potent compound (1) in this study showed MIC(50) value of 2.22 microg/mL against Candida albicans.
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)