Merck
  • Blood and lymphatic systems are segregated by the FLCN tumor suppressor.
Blood and lymphatic systems are segregated by the FLCN tumor suppressor.
Nature communications (2020-12-11)
Ikue Tai-Nagara, Yukiko Hasumi, Dai Kusumoto, Hisashi Hasumi, Keisuke Okabe, Tomofumi Ando, Fumio Matsuzaki, Fumiko Itoh, Hideyuki Saya, Chang Liu, Wenling Li, Yoh-Suke Mukouyama, W Marston Linehan, Xinyi Liu, Masanori Hirashima, Yutaka Suzuki, Shintaro Funasaki, Yorifumi Satou, Mitsuko Furuya, Masaya Baba, Yoshiaki Kubota
ABSTRACT
Blood and lymphatic vessels structurally bear a strong resemblance but never share a lumen, thus maintaining their distinct functions. Although lymphatic vessels initially arise from embryonic veins, the molecular mechanism that maintains separation of these two systems has not been elucidated. Here, we show that genetic deficiency of Folliculin, a tumor suppressor, leads to misconnection of blood and lymphatic vessels in mice and humans. Absence of Folliculin results in the appearance of lymphatic-biased venous endothelial cells caused by ectopic expression of Prox1, a master transcription factor for lymphatic specification. Mechanistically, this phenotype is ascribed to nuclear translocation of the basic helix-loop-helix transcription factor Transcription Factor E3 (TFE3), binding to a regulatory element of Prox1, thereby enhancing its venous expression. Overall, these data demonstrate that Folliculin acts as a gatekeeper that maintains separation of blood and lymphatic vessels by limiting the plasticity of committed endothelial cells.
Sigma-Aldrich
MISSION® esiRNA, targeting human TFE3
Sigma-Aldrich
Anti-PECAM-1 Antibody, clone 2H8, Azide Free, clone 2H8, Chemicon®, from hamster(Armenian)
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution