Merck
  • Nasally Administered Lactococcus lactis Secreting Heme Oxygenase-1 Attenuates Murine Emphysema.
Nasally Administered Lactococcus lactis Secreting Heme Oxygenase-1 Attenuates Murine Emphysema.
Antioxidants (Basel, Switzerland) (2020-10-31)
Kentaro Yumoto, Takashi Sato, Kentaro Nakashima, Fu Namai, Suguru Shigemori, Takeshi Shimosato, Takeshi Kaneko
ABSTRACT
Emphysema, a type of lung-destroying condition associated with chronic obstructive pulmonary disease (COPD), is an inflammatory lung disease mainly due to cigarette smoke exposure. As there is no curative therapy, prevention should be considered first by cessation of smoking to avoid exposure to oxidative stresses and inflammatory mediators. In addition, therapies involving antioxidative and/or anti-inflammatory agents such as heme oxygenase (HO)-1 are candidate treatments. We developed a new tool using genetically modified Lactococcus lactis to deliver recombinant HO-1 to the lungs. Using an elastase-induced emphysema model mimicking COPD, we evaluated the effect of nasally administered L. lactis secreting HO-1 (HO-1 lactis) on cellular and molecular responses in the lungs and further disease progression. Nasally administered HO-1 lactis resulted in (1) overexpression of HO-1 in the lungs and serum and (2) attenuation of emphysema progression evaluated both physiologically and morphologically. There was a transient 5-10% weight loss compared to baseline through trafficking to the lungs when administering 1.0 × 109 cells/mouse; however, this did not impact either survival or final body weight. These results suggest that delivering HO-1 using genetically modified L. lactis through the airways could be a safe and potentially effective therapeutic approach for COPD.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-His-Tag antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM146, purified immunoglobulin