Merck
  • The nongenotoxic carcinogens naphthalene and para-dichlorobenzene suppress apoptosis in Caenorhabditis elegans.
The nongenotoxic carcinogens naphthalene and para-dichlorobenzene suppress apoptosis in Caenorhabditis elegans.
Nature chemical biology (2006-05-16)
David Kokel, Yehua Li, Jun Qin, Ding Xue
ABSTRACT
Naphthalene (1) and para-dichlorobenzene (PDCB, 2), which are widely used as moth repellents and air fresheners, cause cancer in rodents and are potential human carcinogens. However, their mechanisms of action remain unclear. Here we describe a novel method for delivering and screening hydrophobic chemicals in C. elegans and apply this technique to investigate the ways in which naphthalene and PDCB may promote tumorigenesis in mammals. We show that naphthalene and PDCB inhibit apoptosis in C. elegans, a result that suggests a cellular mechanism by which these chemicals may promote the survival and proliferation of latent tumor cells. In addition, we find that a naphthalene metabolite directly inactivates caspases by oxidizing the active site cysteine residue; this suggests a molecular mechanism by which these chemicals suppress apoptosis. Naphthalene and PDCB are the first small-molecule apoptosis inhibitors identified in C. elegans. The power of C. elegans molecular genetics, in combination with the possibility of carrying out large-scale chemical screens in this organism, makes C. elegans an attractive and economic animal model for both toxicological studies and drug screens.
Product Description
Supelco
Melting point standard 79-81°C, analytical standard
Sigma-Aldrich
1,4-Naphthoquinone, purum, ≥96.5% (HPLC)
Sigma-Aldrich
1,4-Naphthoquinone, 97%
Sigma-Aldrich
1,4-Dichlorobenzene, ≥99%
Supelco
Naphthalene solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Supelco
Naphthalene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Bis(2-ethylhexyl) phthalate, Selectophore
Supelco
Bis(2-ethylhexyl) phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dioctyl phthalate, ≥99.5%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
Naphthalene, 99%
Supelco
Naphthalene, analytical standard
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%