Merck
  • Symmetric Arginine Dimethylation Is Selectively Required for mRNA Splicing and the Initiation of Type I and Type III Interferon Signaling.
Symmetric Arginine Dimethylation Is Selectively Required for mRNA Splicing and the Initiation of Type I and Type III Interferon Signaling.
Cell reports (2020-02-13)
Patrick J Metz, Keith A Ching, Tao Xie, Paulina Delgado Cuenca, Sherry Niessen, John H Tatlock, Kristen Jensen-Pergakes, Brion W Murray
ABSTRACT
Alternative splicing is well understood to enhance proteome diversity as cells respond to stimuli. However, mechanistic understanding for how the spliceosome processes precursor messenger RNA (mRNA) transcripts to achieve template diversification is incomplete. We use recently developed enzymatic inhibitors of protein arginine methyltransferase 5 (PRMT5) and human naive T lymphocyte activation as a model system to uncover a precise set of mRNA transcripts that require symmetric arginine dimethylation. This methylation-dependent splicing selectivity is associated with a limited set of signaling pathways that are affected when PRMT5 is inhibited. Specifically, we identify a conserved role for symmetric arginine dimethylation in the induction of antiviral type I and type III interferon signaling following T cell receptor and pattern recognition receptor stimulation in human T lymphocytes and undifferentiated human THP-1 monocytes. Altogether, these findings reveal a mechanism by which cells may be enabled to precisely modulate transcript heterogeneity to orchestrate specific functional outcomes.
Product Description
Millipore
L-Alanine, A non-essential amino acid.
Sigma-Aldrich
IgG1 Isotype Control from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-SmB antibody, Mouse Monoclonal, clone 12F5, purified from hybridoma cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Roche
cOmplete ULTRA Tablets, Mini, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Sigma-Aldrich
ClickTech EdU Cell Proliferation Kit 488 for FC, sufficient for 50 assays