Merck
  • Interference by naturally occurring fatty acids in a noncellular enzyme-based aromatase bioassay.
Interference by naturally occurring fatty acids in a noncellular enzyme-based aromatase bioassay.
Journal of natural products (2006-04-29)
Marcy J Balunas, Bin Su, Serena Landini, Robert W Brueggemeier, A Douglas Kinghorn
ABSTRACT
Natural product drug discovery efforts frequently utilize noncellular screening assays. Fatty acids are commonly found in natural product extracts, and some have been shown to interfere with noncellular assays. Several pure fatty acids were tested using a noncellular aromatase assay, with the unsaturated analogues showing strong inhibitory activity, while the saturated analogues were inactive. Unsaturated fatty acids were further tested against SK-BR-3 hormone-independent human breast cancer cells that overexpress aromatase and were found to be inactive. In natural product screening efforts, especially using plant seeds, it is recommended that extracts active in noncellular bioassays should be dereplicated for the presence of fatty acids prior to bioassay-guided fractionation.
Sigma-Aldrich
Pentadecanoic acid, 99%
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
cis-5,8,11,14,17-Eicosapentaenoic acid, ≥99%
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Oleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Pentadecanoic acid, ~99% (capillary GC)
Sigma-Aldrich
DL-Aminoglutethimide
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
cis-4,7,10,13,16,19-Docosahexaenoic acid, ≥98%
Sigma-Aldrich
Oleic acid, natural, FCC
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Supelco
cis-5,8,11,14,17-Eicosapentaenoic acid, analytical standard
Supelco
Pentadecanoic acid, analytical standard
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Supelco
Oleic acid, Selectophore, for ion-selective electrodes, ≥99%
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
all-cis-7,10,13,16,19-Docosapentaenoic acid, synthetic, ≥97%
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Myristic acid, Sigma Grade, ≥99%
Sigma-Aldrich
Myristic acid, ≥98.0% (GC)
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Supelco
Myristic acid, analytical standard