Merck
  • Vanillic acid glycoside and quinic acid derivatives from Gardeniae Fructus.
Vanillic acid glycoside and quinic acid derivatives from Gardeniae Fructus.
Journal of natural products (2006-04-29)
Hyoung Ja Kim, Eun Jung Kim, Seon Hee Seo, Cha-Gyun Shin, Changbae Jin, Yong Sup Lee
ABSTRACT
Bioassay-directed chromatographic fractionation of an ethyl acetate extract of Gardenia jasminoides (Gardeniae Fructus) afforded a new vanillic acid 4-O-beta-d-(6'-sinapoyl)glucopyranoside (1) and five new quinic acid derivatives, methyl 5-O-caffeoyl-3-O-sinapoylquinate (2), ethyl 5-O-caffeoyl-3-O-sinapoylquinate (3), methyl 5-O-caffeoyl-4-O-sinapoylquinate (4), ethyl 5-O-caffeoyl-4-O-sinapoylquinate (5), and methyl 3,5-di-O-caffeoyl-4-O-(3-hydroxy-3-methyl)glutaroylquinate (6), together with three known quinic acid derivatives, two flavonoids, two iridoids, and two phenolic compounds. The structures of new compounds were elucidated by the aid of spectroscopic methods. These compounds were assessed for antioxidant activity using three different cell-free bioassay systems and for HIV-1 integrase inhibitory activity. Five new quinic acid derivatives showed potent DPPH radical scavenging, superoxide anion scavenging, and lipid peroxidation inhibition activities. These new quinic acid derivatives also exhibited HIV-1 integrase inhibitory activity.
Product Description
Supelco
Resveratrol, analytical standard
Isoquercitrin, primary reference standard
Supelco
Quercetin 3-glucoside, analytical standard
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
(+)-α-Tocopherol, from vegetable oil, Type V, ~1000 IU/g
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
(+)-α-Tocopherol, Type VI, from vegetable oil, liquid (≥0.88M based on potency, density and molecular wt.), BioReagent, suitable for insect cell culture, ≥1000 IU/g
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Quercetin 3-β-D-glucoside, ≥90% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, 99%