Merck
Synthesis and biological activities of nucleoside-estradiol conjugates.
Bioorganic & medicinal chemistry letters (2005-11-09)
Hasrat Ali, Naseem Ahmed, Guillaume Tessier, Johan E van Lier
ABSTRACT
Nucleosides were coupled to estradiol via a 17alpha-ethynyl spacer group using Pd(II) as a catalyst. The conjugates were evaluated in vitro for estrogen receptor (ER) binding affinity and cytotoxicity against cell lines with and without ER. The highest receptor binding affinities (RBA approximately 3) were observed with conjugates coupled via a relative long spacer group, while none of the conjugates exhibited cytotoxicity against either cell lines.
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
β-Estradiol, analytical standard
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Estradiol, meets USP testing specifications