Merck
  • Collagen-rich omentum is a premetastatic niche for integrin α2-mediated peritoneal metastasis.
Collagen-rich omentum is a premetastatic niche for integrin α2-mediated peritoneal metastasis.
eLife (2020-10-08)
Yen-Lin Huang, Ching-Yeu Liang, Danilo Ritz, Ricardo Coelho, Dedy Septiadi, Manuela Estermann, Cécile Cumin, Natalie Rimmer, Andreas Schötzau, Mónica Núñez López, André Fedier, Martina Konantz, Tatjana Vlajnic, Diego Calabrese, Claudia Lengerke, Leonor David, Barbara Rothen-Rutishauser, Francis Jacob, Viola Heinzelmann-Schwarz
ABSTRACT
The extracellular matrix (ECM) plays critical roles in tumor progression and metastasis. However, the contribution of ECM proteins to early metastatic onset in the peritoneal cavity remains unexplored. Here, we suggest a new route of metastasis through the interaction of integrin alpha 2 (ITGA2) with collagens enriched in the tumor coinciding with poor outcome in patients with ovarian cancer. Using multiple gene-edited cell lines and patient-derived samples, we demonstrate that ITGA2 triggers cancer cell adhesion to collagen, promotes cell migration, anoikis resistance, mesothelial clearance, and peritoneal metastasis in vitro and in vivo. Mechanistically, phosphoproteomics identify an ITGA2-dependent phosphorylation of focal adhesion kinase and mitogen-activated protein kinase pathway leading to enhanced oncogenic properties. Consequently, specific inhibition of ITGA2-mediated cancer cell-collagen interaction or targeting focal adhesion signaling may present an opportunity for therapeutic intervention of metastatic spread in ovarian cancer.
Sigma-Aldrich
Anti-COL1A1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-Integrin alpha 10 (ITGA10) Antibody, from rabbit, purified by affinity chromatography