Merck
  • Enrichment of histones from patient samples for mass spectrometry-based analysis of post-translational modifications.
Enrichment of histones from patient samples for mass spectrometry-based analysis of post-translational modifications.
Methods (San Diego, Calif.) (2019-10-13)
Roberta Noberini, Camilla Restellini, Evelyn Oliva Savoia, Tiziana Bonaldi
ABSTRACT
Aberrations in histone post-translational modifications (PTMs) have been implicated with the development of numerous pathologies, including cancer. Therefore, profiling histone PTMs in patient samples could provide information useful for the identification of epigenetic biomarkers, as well as for the discovery of potential novel targets. While antibody-based methods have been traditionally employed to analyze histone PTM in clinical samples, mass spectrometry (MS) can provide a more comprehensive, unbiased and quantitative view on histones and their PTMs. To combine the power of MS-based methods and the potential offered by histone PTM profiling of clinical samples, we have recently developed a series of methods for the extraction and enrichment of histones from different types of patient samples, including formalin-fixed paraffin-embedded tissues, fresh- and optimal cutting temperature-frozen tissues, and primary cells. Here, we provide a detailed description of these protocols, together with indications on the expected results and the most suitable workflow to be used downstream of each procedure.
Product Description
Supelco
Empore SPE Disks, matrix active group C18, diam. 47 mm, pk of 20
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Trizma® hydrochloride, BioPerformance Certified, suitable for cell culture, ≥99.0% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
Leupeptin, synthetic, ≥85% (HPLC)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)