Merck
Sulfonamide chalcone as a new class of alpha-glucosidase inhibitors.
Bioorganic & medicinal chemistry letters (2005-10-06)
Woo Duck Seo, Jin Hyo Kim, Jae Eun Kang, Hyung Won Ryu, Marcus J Curtis-Long, Hyun Sun Lee, Min Suk Yang, Ki Hun Park
ABSTRACT
Chalcones 1-20, a new class of glycosidase inhibitors, were synthesized, and their glycosidase inhibitory activities were investigated. Non-aminochalcones 1-12 had no inhibitory activity, however, aminochalcones 13-20 had strong glycosidase (alpha-glucosidase, alpha-amylase, and beta-amylase) inhibitory activities. In particular, sulfonamide chalcones 17-20 had more potent alpha-glucosidase inhibitory activity than aminated chalcone 13-16. 4'-(p-Toluenesulfonamide)-3,4-dihydroxy chalcone 20 (IC(50)=0.4microM) was the best inhibitor against alpha-glucosidase, and these sulfonamide chalcones showed non-competitive inhibition.
Sigma-Aldrich
Myricetin, ≥96.0%, crystalline
Sigma-Aldrich
Myricetin, ≥96.0% (HPLC)
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea