Merck
  • Age-Related Dopaminergic Innervation Augments T Helper 2-Type Allergic Inflammation in the Postnatal Lung.
Age-Related Dopaminergic Innervation Augments T Helper 2-Type Allergic Inflammation in the Postnatal Lung.
Immunity (2019-11-24)
Wei Wang, Jonathan A Cohen, Antonia Wallrapp, Kenneth G Trieu, Juliana Barrios, Fengzhi Shao, Nandini Krishnamoorthy, Vijay K Kuchroo, Matthew R Jones, Alan Fine, Yan Bai, Xingbin Ai
ABSTRACT
Young children are more susceptible to developing allergic asthma than adults. As neural innervation of the peripheral tissue continues to develop after birth, neurons may modulate tissue inflammation in an age-related manner. Here we showed that sympathetic nerves underwent a dopaminergic-to-adrenergic transition during post-natal development of the lung in mice and humans. Dopamine signaled through a specific dopamine receptor (DRD4) to promote T helper 2 (Th2) cell differentiation. The dopamine-DRD4 pathway acted synergistically with the cytokine IL-4 by upregulating IL-2-STAT5 signaling and reducing inhibitory histone trimethylation at Th2 gene loci. In murine models of allergen exposure, the dopamine-DRD4 pathway augmented Th2 inflammation in the lungs of young mice. However, this pathway operated marginally after sympathetic nerves became adrenergic in the adult lung. Taken together, the communication between dopaminergic nerves and CD4+ T cells provides an age-related mechanism underlying the susceptibility to allergic inflammation in the early lung.
Sigma-Aldrich
Anti-Calcitonin Gene Related Peptide antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Acetyl-β-methylcholine chloride, ≥98% (TLC), powder
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
Histone Methyltransferase EZH2 Inhibitor, DZNep, Histone Methyltransferase EZH2 Inhibitor, DZNep, CAS 120964-45-6, is a cell-permeable inhibitor of EZH2-mediated trimethylation of K27 on histone H3.
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Dopamine β Hydroxylase Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-CD11b Antibody (human/mouse), APC, clone M1/70, clone M1/70, 0.2 mg/mL, from rat
Sigma-Aldrich
Anti-DRD4 Antibody, clone 2B9, clone 2B9, from mouse