Merck
  • Intraflagellar Transport 80 Is Required for Cilia Construction and Maintenance in Paramecium tetraurelia.
Intraflagellar Transport 80 Is Required for Cilia Construction and Maintenance in Paramecium tetraurelia.
The Journal of eukaryotic microbiology (2020-05-06)
Lei Shi, Yuhao Chi, Xiangyu Shen, Guoliang Lu, Yuan Shen
ABSTRACT
Intraflagellar transport (IFT) represents a bidirectional dynamic process that carries cargo essential for cilia building and the maintenance of ciliary function, which is important for the locomotion of single cells, intracellular and intercellular signalling transduction. Accumulated evidence has revealed that defects in IFT cause several clinical disorders. Here, we determined the role of IFT80, an IFT-B protein that is mutated in Jeune asphyxiating thoracic dystrophy. Using the RNAi method in the ciliate Paramecium as model, we found that loss of IFT80 prevents cilia biogenesis and causes strong cell lethality. A specific antibody against IFT80 was also prepared in our study, which labelled IFT80 in cilia of Paramecium. GFP fusion experiments were performed to illustrate the dynamic movement of IFT-A and IFT-B proteins in cilia of Paramecium; then, we found that the depletion of IFT80 in cells prevents IFT-A and IFT-B proteins from entering the cilia. Our results showed the distribution change of other IFT proteins in cells that were depleted of IFT80, and we discuss the possible roles of IFT80 in Paramecium.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−TRITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Hoechst 33258 solution, 1 mg/mL in H2O, ≥98.0% (HPLC)
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
Anti-beta-Actin antibody, Rabbit monoclonal, clone SP124, recombinant, expressed in proprietary host, affinity isolated antibody