Merck
  • Art-on-a-Chip: Preserving Microfluidic Chips for Visualization and Permanent Display.
Art-on-a-Chip: Preserving Microfluidic Chips for Visualization and Permanent Display.
Small (Weinheim an der Bergstrasse, Germany) (2020-07-24)
Rebecca Soffe, Albert J Mach, Sevgi Onal, Volker Nock, Luke P Lee, J Tanner Nevill
ABSTRACT
"After a certain high level of technical skill is achieved, science and art tend to coalesce in aesthetics, plasticity, and form. The greatest scientists are always artists as well." said Albert Einstein. Currently, photographic images bridge the gap between microfluidic/lab-on-a-chip devices and art. However, the microfluidic chip itself should be a form of art. Here, novel vibrant epoxy dyes are presented in combination with a simple process to fill and preserve microfluidic chips, to produce microfluidic art or art-on-a-chip. In addition, this process can be used to produce epoxy dye patterned substrates that preserve the geometry of the microfluidic channels-height within 10% of the mold master. This simple approach for preserving microfluidic chips with vibrant, colorful, and long-lasting epoxy dyes creates microfluidic chips that can easily be visualized and photographed repeatedly, for at least 11 years, and hence enabling researchers to showcase their microfluidic chips to potential graduate students, investors, and collaborators.
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Sudan I, Dye content ≥95 %
Sigma-Aldrich
Scienceware® vacuum desiccator, flange O.D. × I.D. 230 mm × 197 mm
Sigma-Aldrich
Trichloro(1H,1H,2H,2H-perfluorooctyl)silane, 97%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%