Merck
  • Neuroprotective potential of antihyperglycemic drug metformin in streptozocin-induced rat model of sporadic Alzheimer's disease.
Neuroprotective potential of antihyperglycemic drug metformin in streptozocin-induced rat model of sporadic Alzheimer's disease.
European journal of pharmacology (2020-06-25)
Vladimirs Pilipenko, Karīna Narbute, Jolanta Pupure, Ilva Kristiāna Langrate, Ruta Muceniece, Vija Kluša
ABSTRACT
The earliest hallmarks of sporadic Alzheimer's disease (sAD) are impaired glucose metabolism, chronic neuroinflammation, diminished synaptic plasticity and subsequent cognitive decline. The safest antidiabetic drug metformin has shown both glucose metabolism-improving and cognition-enhancing action in type 2 diabetes patients and diabetic model animals. However, metformin has not been previously studied in intracerebroventricular streptozocin (STZ)-induced model of sAD. Therefore, our aim was to assess the preventive action of metformin in sAD model-rats. Firstly, the actions of metformin (75 and 100 mg/kg) on cognitive functions and sociability were examined. Secondly, we wanted to identify whether behavioral effects of metformin were provided via its action on brain glucose transport, neuronal/glial uptake and metabolism. Thirdly, the effects of metformin on neuroinflammation, acetylcholine esterase density and activity, as well as on synaptic plasticity were determined. Our results showed that metformin reversed STZ-induced impairments in spatial learning/memory performance and sociability, coinciding with normalization of brain glucose transport, uptake and metabolism. Microgliosis and astrogliosis were ameliorated by metformin in sAD model rats. Metformin also preserved hippocampal synaptic plasticity and normalized acetylcholine cleavage in the cortical and hippocampal tissues, as well as inhibited acetylcholine esterase activity in vitro. These data indicate the promise of further research of metformin in early brain pathologies to stop neurodegenerative before severe cognitive decline occurs.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
Ethopropazine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
RIPA Buffer