Merck
  • Reducing NADPH Synthesis Counteracts Diabetic Nephropathy through Restoration of AMPK Activity in Type 1 Diabetic Rats.
Reducing NADPH Synthesis Counteracts Diabetic Nephropathy through Restoration of AMPK Activity in Type 1 Diabetic Rats.
Cell reports (2020-10-01)
Xiao Wei, Zongshi Lu, Li Li, Hexuan Zhang, Fang Sun, Huan Ma, Lijuan Wang, Yingru Hu, Zhencheng Yan, Hongting Zheng, Gangyi Yang, Daoyan Liu, Martin Tepel, Peng Gao, Zhiming Zhu
ABSTRACT
Diabetic nephropathy (DN) is a major complication of diabetes mellitus and a primary cause of end-stage renal failure. Clinical studies indicate that metabolic surgery improves DN; however, the mechanism remains unclear. Here, we report that Roux-en-Y Gastric Bypass (RYGB) surgery significantly blocked and reversed DN without affecting the insulin signaling pathway. This protective role of RYGB surgery is almost blocked by either inhibition or knockout of 5'AMP-activated protein kinase (AMPK) in podocytes. Furthermore, mRNA microarray data reveal that RYGB surgery obviously reduced the gene expression involved in nicotinamide adenine dinucleotide phosphate (NAPDH) synthesis. The expression of a key NADPH synthase, hexose-6-phosphate dehydrogenase (H6PD), was inhibited by the low plasma corticosterone level after surgery. In addition, blocking NAPDH synthesis by knocking down H6PD mimicked the beneficial role of RYGB surgery through activation of AMPK in podocytes. Therefore, this study demonstrates that reducing NADPH production is critical for renal AMPK activation in response to RYGB surgery.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
C646, ≥98% (HPLC)
Sigma-Aldrich
Sodium succinate dibasic hexahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide hydrate, ≥99%
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)