Merck
  • Stability and anti-topoisomerase activity of phenolic compounds of Capsicum annuum "Serrano" after gastrointestinal digestion and in vitro colonic fermentation.
Stability and anti-topoisomerase activity of phenolic compounds of Capsicum annuum "Serrano" after gastrointestinal digestion and in vitro colonic fermentation.
International journal of food sciences and nutrition (2020-03-07)
Alicia P Cárdenas-Castro, Emilio Alvarez-Parrilla, Efigenia Montalvo-González, Jorge A Sánchez-Burgos, Koen Venema, Sonia G Sáyago-Ayerdi
ABSTRACT
"Serrano" pepper is extensively used in Mexican cuisine. The aim of this study was to identify the bioaccessible phenolic compounds (PC) of "Serrano" pepper as well as short-chain fatty acids (SCFA) produced and PC bioconverted using an in vitro step-wise gastromimetic model of the intestinal digestion and anaerobic fermentation of the isolated indigestible fraction (IF). The anti-topoisomerase activity of the fermented samples was also evaluated. PC bioaccessibility was about 45% in the small intestine. Chlorogenic acid and capsaicin were identified during the intestinal digestion, while quercetin was identified as available to the gut microbiota. After 48-h fermentation, SCFA molar ratio was 77:11:12 for acetic, propionic and butyric acid. The PC identified in IF and after 12 h of fermentation showed anti-topoisomerase activity. A synergistic effect among the PC and gut metabolites mixture was observed, which indicates a possible antiproliferative mechanism that should be tested in further studies.
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Ellagic acid, ≥95% (HPLC), powder, from tree bark
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea
Sigma-Aldrich
Vanillic acid, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Bile extract porcine
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
Sigma-Aldrich
Dihydrocapsaicin, from Capsicum sp., ≥85%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%