Merck
  • Silencing of miR490-3p by H. pylori activates DARPP-32 and induces resistance to gefitinib.
Silencing of miR490-3p by H. pylori activates DARPP-32 and induces resistance to gefitinib.
Cancer letters (2020-08-01)
Shoumin Zhu, Shayan Khalafi, Zheng Chen, Julio Poveda, Dunfa Peng, Heng Lu, Mohammed Soutto, Jianwen Que, Monica Garcia-Buitrago, Alexander Zaika, Wael El-Rifai
ABSTRACT
Infection with Helicobacter pylori (H. pylori) is the main risk factor for gastric carcinogenesis. In this study, we investigated the expression, molecular functions, and downstream effectors of miR490-3p in gastric cancer. We used in vitro and in vivo models to investigate the role of H. pylori in regulating miR490-3p, DARPP-32-dependent functions, and therapeutic resistance. Human and mouse neoplastic gastric lesions demonstrated a negative correlation between DARPP-32 and miR490-3p expression (R = -0.58, P < 0.01). This was also detected following infection with H. pylori (R = -0.66, P < 0.01). Molecular assays confirmed DARPP-32 as a direct target of miR490-3p. CHRM2, the host gene of miR490-3p, was hypermethylated and downregulated in neoplastic gastric tissues (P < 0.05). H. pylori induced methylation and downregulation of CHRM2 and miR490-3p. Functionally, the reconstitution of miR490-3p sensitized cancer cells to gefitinib by inactivating DRAPP-32-dependent AKT and STAT3 pathways. Patients with low miR490-3p or high DARPP-32 expression had decreased overall survival (P < 0.05). Hypermethylation-mediated silencing of CHRM2 and miR490-3p by H. pylori increased DARPP-32 expression. Downregulation of miR490-3p in gastric cancer plays a role in gefitinib response by inducing DARPP-32-mediated activation of PI3K/AKT, STAT3 signaling pathways.
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
MISSION® esiRNA, targeting human PPP1R1B