- Claudin 4 expression in triple-negative breast cancer: correlation with androgen receptors and Ki-67 expression.
Claudin 4 expression in triple-negative breast cancer: correlation with androgen receptors and Ki-67 expression.
Breast cancer is the most common malignancy in women and the leading cause of cancer mortality worldwide. Triple-negative breast cancer (TNBC) is an important phenotype of breast cancer that accounts for a relatively small number of breast cancer cases but still represent a focus of increasing interest at the clinical, biological, and epidemiological level. Claudins are the major component of the tight junction, and only a few studies have addressed the role of claudins in breast cancer, especially TNBC. Androgen receptors (ARs), as members of the nuclear receptor superfamily, are known to be involved in a complex network of signaling pathways that collectively regulate cell proliferation. However, roles of AR in breast cancer development and progression have not been very clearly understood. The proliferation marker Ki-67 has been confirmed as an independent predictive and prognostic factor in early breast cancer. The aims of this study are to identify the clinicopathologic associations and prognostic value of claudin 4 expression in TNBC and to correlate claudin 4 expression with AR status and Ki-67 expression. Paraffin blocks obtained from 56 female patients with triple-negative primary invasive ductal breast carcinomas were analyzed for claudin 4, AR, and Ki-67 immunohistochemical expression. High levels of claudin 4 expression were detected in 66.1% of TNBC cases. There was a significant positive correlation with age, tumor size, grade, nodal status, metastasis, and Ki-67 expression (all P < .05) and negative correlation with AR status (P < .001). Androgen receptor showed positivity in 29 cases (51.78%). There was a statistical negative correlation with the all the studied clinicopathologic parameters, claudin 4 and Ki-67 expression. High claudin 4 expression, negative AR expression, and high Ki-67 index would provide a strong prognostic power to differentiate the patients with worse outcome among TNBC patients. Moreover, target treatment for TNBC cells expressing claudin 4 or AR enriched would be valuable for future therapies.