Merck
  • Multiplex Point-of-Care Tests for the Determination of Antibodies after Acellular Pertussis Vaccination.
Multiplex Point-of-Care Tests for the Determination of Antibodies after Acellular Pertussis Vaccination.
Diagnostics (Basel, Switzerland) (2020-04-02)
Aapo Knuutila, Carita Rautanen, Jussi Mertsola, Qiushui He
ABSTRACT
Most of the current serological diagnosis of pertussis is based on pertussis toxin (PT) IgG antibodies and does not differentiate between vaccination and infection-induced antibodies. PT is included in all of acellular pertussis vaccines available in the world. Multiplex testing of non-vaccine antigen-related antibodies has the potential to improve the diagnostic outcome of these assays. In this study, we developed a quantitatively spatial multiplex lateral flow immunoassay (LFIA) for the detection of IgG antibodies directed against PT, pertactin (PRN), and filamentous hemagglutinin (FHA). The assay was evaluated with serum samples with varying anti-PT, anti-PRN, and anti-FHA IgG levels and the result was compared to those obtained with standardized ELISA. The developed assay showed good specificity with PT and PRN antibodies and semiquantification throughout the antigen combinations. This exploratory study indicates that the multiplex LFIA is specific and sensitive, and a similar test platform with alternative antigens could be suitable for new type of pertussis serology.
Product Description
Millipore
Cellulose Absorbent Sample Pads, thickness 0.83 mm, filter L × W 20 cm × 30 cm, pack of 100 sheets