Merck
  • Naphthalimide-Based Aggregation-Induced Emissive Polymeric Hydrogels for Fluorescent Pattern Switch and Biomimetic Actuators.
Naphthalimide-Based Aggregation-Induced Emissive Polymeric Hydrogels for Fluorescent Pattern Switch and Biomimetic Actuators.
Macromolecular rapid communications (2020-05-14)
Hao Liu, Shuxin Wei, Huiyu Qiu, Beibei Zhan, Qingquan Liu, Wei Lu, Jiawei Zhang, To Ngai, Tao Chen
ABSTRACT
Substituted naphthalimide (NI) moieties are highly versatile and newly recognized aggregation-induced emission (AIE) building blocks for many potentially useful smart molecules, polymers, and nanoparticles. However, the introduction of NI fluorophore into cross-linked polymeric networks to prepare AIE-active hydrogels still remains underdeveloped. Herein, a novel naphthalimide-based aggregation-induced emissive polymeric hydrogel is reported, followed by its proof-of-concept applications as fluorescence pattern switch and biomimetic actuator. The hydrogel, bearing semi-interpenetrating polymer networks, is synthesized starting from N-isopropylacrylamide, hydroxyethyl methacrylate, and a newly designed NI monomer (4-phenoxy-N-allyl-1,8-naphthalimide, PhAN). Rational molecular design for AIE-active PhAN monomer lies in modification of the NI core with rigid and bulky phenoxy group to break its planarity to produce desirable propeller-shaped molecular conformation. The as-prepared hydrogel is proved to be a aggregation-induced blue-light-emitting hydrogel. It also shows volume phase transition behavior and is endowed with thermally responsive synergistic emission and transmittance change, thus enabling simultaneous regulation of two optical properties merely by one single stimulus. These useful advantages further encourage fabrication of several proto-type fluorescence pattern switching and biomimetic actuating devices. This study may not only enlarge the list of fluorescent hydrogels but also serve as a novel smart optical platform for potential anticounterfeiting, sensing, displaying, or actuating applications.
Sigma-Aldrich
4-Bromo-1,8-naphthalic anhydride, 95%
Pricing and availability is not currently available.