Merck
  • Cancer-associated fibroblasts-derived gamma-glutamyltransferase 5 promotes tumor growth and drug resistance in lung adenocarcinoma.
Cancer-associated fibroblasts-derived gamma-glutamyltransferase 5 promotes tumor growth and drug resistance in lung adenocarcinoma.
Aging (2020-07-09)
Jia-Ru Wei, Jun Dong, Lei Li
ABSTRACT
Gamma-glutamyltransferase 5 (GGT5) is a member of the gamma-glutamyl transpeptidase gene family with the capacity of cleaving the gamma-glutamyl moiety of glutathione, but its role in cancer progression has never been revealed. In this study, we found that gene GGT5 was highly expressed in cancer-associated fibroblasts (CAFs) in lung adenocarcinoma, predicting the unfavorable survival of patients with lung adenocarcinoma. Cell growth, foci formation and spheres formation analyses showed that cancer cell proliferation was attenuated under treatment with the conditioned media from GGT5-silenced CAFs. Moreover, high expression of GGT5 in CAFs enhanced the drug resistance of cancer cells by increasing intracellular glutathione and reducing the intracellular reactive oxygen species in cancer cells. In mouse xenograft model, we proved that targeting GGT5 with a small-molecule inhibitor GGsTop could inhibit tumor growth and increase the chemosensitivity of cancer cells. Taken together, our study illuminates that high level of GGT5 in CAFs contributes to cancer cell survival and drug resistance, indicating that GGT5 may be a promising therapeutic target in lung adenocarcinoma.