Merck
  • Elevated COUP-TFII expression in dopaminergic neurons accelerates the progression of Parkinson's disease through mitochondrial dysfunction.
Elevated COUP-TFII expression in dopaminergic neurons accelerates the progression of Parkinson's disease through mitochondrial dysfunction.
PLoS genetics (2020-06-25)
Chung-Yang Kao, Mafei Xu, Leiming Wang, Shih-Chieh Lin, Hui-Ju Lee, Lita Duraine, Hugo J Bellen, David S Goldstein, Sophia Y Tsai, Ming-Jer Tsai
ABSTRACT
Parkinson's disease (PD) is a neurodegenerative disorder featuring progressive loss of midbrain dopaminergic (DA) neurons that leads to motor symptoms. The etiology and pathogenesis of PD are not clear. We found that expression of COUP-TFII, an orphan nuclear receptor, in DA neurons is upregulated in PD patients through the analysis of public datasets. We show here that through epigenetic regulation, COUP-TFII contributes to oxidative stress, suggesting that COUP-TFII may play a role in PD pathogenesis. Elevated COUP-TFII expression specifically in DA neurons evokes DA neuronal loss in mice and accelerates the progression of phenotypes in a PD mouse model, MitoPark. Compared to control mice, those with elevated COUP-TFII expression displayed reduced cristae in mitochondria and enhanced cellular electron-dense vacuoles in the substantia nigra pars compacta. Mechanistically, we found that overexpression of COUP-TFII disturbs mitochondrial pathways, resulting in mitochondrial dysfunction. In particular, there is repressed expression of genes encoding cytosolic aldehyde dehydrogenases, which could enhance oxidative stress and interfere with mitochondrial function via 3,4-dihydroxyphenylacetaldehyde (DOPAL) buildup in DA neurons. Importantly, under-expression of COUP-TFII in DA neurons slowed the deterioration in motor functions of MitoPark mice. Taken together, our results suggest that COUP-TFII may be an important contributor to PD development and a potential therapeutic target.
Product Description
Supelco
Paraquat dichloride hydrate, PESTANAL®, analytical standard
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
D-(+)-Trehalose dihydrate, from Saccharomyces cerevisiae, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Roche
First Strand cDNA Synthesis Kit for RT-PCR (AMV), sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
L-3,4-Dihydroxyphenylalanine methyl ester hydrochloride, solid
Sigma-Aldrich
Benserazide hydrochloride, ≥98% (HPLC), solid
Sigma-Aldrich
5-Aza-2′-deoxycytidine, ≥97%
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Penicillin–Streptomycin Solution Hybri-Max, with 10,000 units penicillin and 10mg/ml streptomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for hybridoma
Sigma-Aldrich
Oligomycin from Streptomyces diastatochromogenes, ≥90% total oligomycins basis (HPLC)
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Rabbit pAb, liquid, Calbiochem®
Sigma-Aldrich
Anti-Dopamine Transporter Antibody, NT, clone DAT-Nt, culture supernatant, clone DAT-Nt, Chemicon®