Merck
  • Treatment with 2-methoxyestradiol increases endothelial nitric oxide synthase activity via scavenger receptor class BI in human umbilical vein endothelial cells.
Treatment with 2-methoxyestradiol increases endothelial nitric oxide synthase activity via scavenger receptor class BI in human umbilical vein endothelial cells.
Molecular human reproduction (2020-04-26)
Tao Dong, Seisuke Sato, Jingya Lyu, Hitomi Imachi, Toshihiro Kobayashi, Kensaku Fukunaga, Takanobu Saheki, Hisakazu Iwama, Guoxing Zhang, Koji Murao
ABSTRACT
Concentrations of 2-methoxyestradiol (2ME2), a principal metabolite of estradiol, are significantly lower in women with severe preeclampsia. Nitric oxide (NO) released by endothelial nitric oxide synthase (eNOS) plays an important role in regulating cardiovascular homeostasis. Importantly, high-density lipoprotein (HDL) stimulates eNOS activity via endothelial human scavenger receptor class B type I (hSR-BI/CLA-1). Here, we aimed to determine the effect of 2ME2 on hSR-BI/CLA-1 expression in human umbilical vein endothelial cells (HUVECs). hSR-BI/CLA-1 expression was measured by real-time PCR, western blotting and reporter gene assays; eNOS activity was assessed by the measurement of eNOS phosphorylation. Both the mRNA and protein concentrations of hSR-BI/CLA-1 were significantly increased by 2ME2 in HUVECs. 2ME2 also dose-dependently increased the transcriptional activity of the hSR-BI/CLA-1 promoter. The effect of 2ME2 treatment on the promoter activity of hSR-BI/CLA-1 was abrogated by treatment with LY294002, a specific inhibitor of phosphatidylinositol 3-kinase, as was the increase in HDL-induced eNOS activation. Notably, constitutively active Akt increased the activity of the hSR-BI/CLA-1 promoter, whereas dominant-negative Akt abolished the effect of 2ME2 treatment on hSR-BI/CLA-1 promoter activity. The nuclear Sp1 protein concentration was significantly increased by exposure to 2ME2 and Sp1 overexpression increased the promoter activity of the hSR-BI/CLA gene. Furthermore, knockdown of Sp1 inhibited the effect of 2ME2 treatment on hSR-BI/CLA-1 protein expression. These results indicate that 2ME2 treatment increases HDL-dependent eNOS phosphorylation by upregulating endothelial hSR-BI/CLA-1 expression, suggesting that 2ME2 has a potential therapeutic value in the treatment of preeclampsia.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture