Merck
  • Polyimide/Ionic Liquid Composite Membranes for Middle and High Temperature Fuel Cell Application: Water Sorption Behavior and Proton Conductivity.
Polyimide/Ionic Liquid Composite Membranes for Middle and High Temperature Fuel Cell Application: Water Sorption Behavior and Proton Conductivity.
Membranes (2020-05-02)
Kateryna Fatyeyeva, Sergiy Rogalsky, Stanislav Makhno, Oksana Tarasyuk, Jorge Arturo Soto Puente, Stéphane Marais
ABSTRACT
Four water insoluble room-temperature protic ionic liquids (PILs) based on the N-alkylimidazolium cation with the alkyl chain length from 1 to 4 and bis(trifluoromethylsulfonyl)imide anion were synthesized and their chemical structure was confirmed by the 1H NMR and 19F NMR analysis. PILs were revealed to be thermally stable up to 360 and 400 °C. At the same time, the proton conductivity of PILs was found to be dependent mostly on the temperature and, to a less extent, on the type of the cation, i.e., the increase of the conductivity from ~3 × 10-4 S/cm at 25 °C to 2 × 10-2 S/cm at 150 °C was observed. The water vapour sorption capacity of PILs was evaluated as a function of relative humidity and the influence of the alkyl chain length on the phase behaviour in the PIL-water system was discussed. The composite polyimide/PILs membranes were prepared by the PIL immobilization in the porous polymer (Matrimid® 5218) film. The composite membranes showed a high level of proton conductivity (~10-3 S/cm) at elevated temperatures (up to 160 °C). The obtained results reveal that the elaborated composite polyimide/PIL membranes are promising candidates for the application as proton exchange membrane at middle and high temperatures.
Sigma-Aldrich
1-Bromopropane, 99%
Sigma-Aldrich
Bromoethane, reagent grade, 98%
Sigma-Aldrich
Sodium hydride, 60 % dispersion in mineral oil
Sigma-Aldrich
Iodomethane, purum, ≥99.0% (GC)