Merck
  • The Profile of Immunoglobulin A and Immunoglobulin G Subclasses in Sprague Dawley Rats Experimentally Infected with Brucella abortus Biotype 1.
The Profile of Immunoglobulin A and Immunoglobulin G Subclasses in Sprague Dawley Rats Experimentally Infected with Brucella abortus Biotype 1.
Vector borne and zoonotic diseases (Larchmont, N.Y.) (2020-02-25)
Mst Minara Khatun, Md Ariful Islam, Byeong Kirl Baek
ABSTRACT
This study measured total serum immunoglobulin A (IgA), immunoglobulin G (IgG)1, IgG2a response against whole cell antigen (WCA), outer membrane protein (OMP), periplasmic protein (PP), cytoplasmic protein (CP), and crude Brucella protein (CBP) of Brucella abortus in experimental brucellosis induced with B. abortus biotype 1 in Sprague Dawley (SD) rats during a 17-week infection period. Six- to 8-week-old SD rats (n = 44) were experimentally infected with 1 × 109 colony forming unit of B. abortus biotype 1 through the intraperitoneal route. Serial serum samples were collected from the rat at 0, 3, 7, 14, 21, 28, 35, 42, 60, 90, and 120 days after inoculation. The sera were tested by enzyme linked immunosorbent assay. We have noticed a very low level and short persistence of IgA antibody in our experiment. The low level and short persistence of IgA antibody suggest that this antibody isotype might not be protective against brucellosis in rats. Both Th1 and Th2 specific immune responses were recorded in our study with the production of IgG1 and IgG2a antibody isotopes, respectively. We noticed significant dominant IgG2a antibody responses over IgG1 responses throughout the experiment (p < 0.001) against WCA and OMP. The mixed Th1 and Th2 dominant immune responses mediated by IgG2a and IgG1 antibody isotypes were observed against CP, PP, and CBP. Data of our study suggest that IgG2a dominant responses in the early stages of disease play the main role in conferring protection against brucellosis and with the progress of disease IgG1 dominant responses were elicited.
Sigma-Aldrich
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)