Merck
  • PHF7 Modulates BRDT Stability and Histone-to-Protamine Exchange during Spermiogenesis.
PHF7 Modulates BRDT Stability and Histone-to-Protamine Exchange during Spermiogenesis.
Cell reports (2020-07-30)
Chang Rok Kim, Taichi Noda, Hyunkyung Kim, Gibeom Kim, Seongwan Park, Yongwoo Na, Seiya Oura, Keisuke Shimada, Injin Bang, Jun-Yeong Ahn, Yong Ryoul Kim, Se Kyu Oh, Hee-Jung Choi, Jong-Seo Kim, Inkyung Jung, Ho Lee, Yuki Okada, Masahito Ikawa, Sung Hee Baek
ABSTRACT
Spermatogenesis is a complex process of sperm generation, including mitosis, meiosis, and spermiogenesis. During spermiogenesis, histones in post-meiotic spermatids are removed from chromatin and replaced by protamines. Although histone-to-protamine exchange is important for sperm nuclear condensation, the underlying regulatory mechanism is still poorly understood. Here, we identify PHD finger protein 7 (PHF7) as an E3 ubiquitin ligase for histone H3K14 in post-meiotic spermatids. Generation of Phf7-deficient mice and Phf7 C160A knockin mice with impaired E3 ubiquitin ligase activity reveals defects in histone-to-protamine exchange caused by dysregulation of histone removal factor Bromodomain, testis-specific (BRDT) in early condensing spermatids. Surprisingly, E3 ubiquitin ligase activity of PHF7 on histone ubiquitination leads to stabilization of BRDT by attenuating ubiquitination of BRDT. Collectively, our findings identify PHF7 as a critical factor for sperm chromatin condensation and contribute to mechanistic understanding of fundamental phenomenon of histone-to-protamine exchange and potential for drug development for the male reproduction system.
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Sigma-Aldrich
Anti-PHF7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-BRDT Antibody, from rabbit, purified by affinity chromatography