Merck
  • The reaction of cucumber to the introduction of ionic liquids into the soil.
The reaction of cucumber to the introduction of ionic liquids into the soil.
Environmental science and pollution research international (2020-06-20)
Robert Biczak, Barbara Pawłowska, Cezary Podsiadło, Martyna Śnioszek, Arkadiusz Telesiński
ABSTRACT
This paper presents the influence of two bromides, tetrabutylammonium and tetrabutylphosphonium, on the growth and development of cucumber seedlings. The tests were performed at two dates, i.e. 10 and 20 days, after the introduction of increasing amounts of ionic liquids (ILs) into the soil. The applied ILs showed phytotoxicity dependent mainly on the concentration of the substance, which is proved by the inhibition of the length of aboveground parts and their roots and the yield of cucumber fresh mass, from which EC50 values were calculated. The phytotoxicity symptoms were the result of oxidative stress, one of the manifestations of which was a decrease in assimilative pigments, linearly correlated with an increase in bromide concentration in the medium. The stress is also proven by the large increase in hydrogen peroxide, malondialdehyde and free proline in cucumber leaves. The reaction of this plant to oxidative stress was an increase in the activity of antioxidative enzymes such as catalase and peroxidase. As a result of statistical analysis, it was proved that all changes of biomarkers of phytotoxicity of examined ILs and oxidative stress indicators in cucumber seedlings depended more on the applied concentration of these salts than on the date of the study.
Sigma-Aldrich
Tetrabutylphosphonium bromide, 98%