Merck
  • Anti-Hypochlorite, Antioxidant, and Catalytic Activity of Three Polyphenol-Rich Super-Foods Investigated with the Use of Coumarin-Based Sensors.
Anti-Hypochlorite, Antioxidant, and Catalytic Activity of Three Polyphenol-Rich Super-Foods Investigated with the Use of Coumarin-Based Sensors.
Biomolecules (2020-05-10)
Karolina Starzak, Tomasz Świergosz, Arkadiusz Matwijczuk, Bernadette Creaven, Janusz Podleśny, Dariusz Karcz
ABSTRACT
The anti-hypochlorite activity of açaí (Euterpe oleracea Mart.), goji (Lyciumbarbarum L.) and schisandra (Schisandrachinensis) fruit extracts were assessed by determining the reactive chlorine species (RCS)-scavenging ability of these three "super-food" berries. In addition, the aqueous extracts obtained were employed as both the media and the catalyst in a green chemistry approach to the synthesis of a coumarin-based fluorescence turn-off sensor, which was then used for anti-hypochlorite activity testing. The aqueous extracts were also assessed for total phenolic content (TPC), using the Folin-Ciocalteu method, and the antioxidant activity using the ABTS+• assay. Moreover, the main water-soluble polyphenolic constituents of the extracts were identified by the HPLC-PDA-ESI-MS technique. Among the extracts tested, açaí demonstrated the highest anti-hypochlorite and antioxidant activities, while the highest TPC value was found for the goji extract. All extracts demonstrated modest catalytic activity as Knoevenagel condensation catalysts.
Sigma-Aldrich
4-(Diethylamino)salicylaldehyde, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
(±)-6-Hydroxy-2,5,7,8-tetramethylchromane-2-carboxylic acid, 97%
Pricing and availability is not currently available.
Rutin, phyproof® Reference Substance
Pricing and availability is not currently available.