Merck
  • Recombinant Lactococcus lactis NZ3900 expressing bioactive human FGF21 reduced body weight of Db/Db mice through the activity of brown adipose tissue.
Recombinant Lactococcus lactis NZ3900 expressing bioactive human FGF21 reduced body weight of Db/Db mice through the activity of brown adipose tissue.
Beneficial microbes (2020-02-19)
W-Y Cao, M Dong, Z-Y Hu, J Wu, Y-C Li, H-D Xu
ABSTRACT
Fibroblast growth factor 21 (FGF21), a metabolism regulator, has an important effect on metabolic diseases, such as obesity and diabetes. It is also expressed in mice, and the murine source has high homology with human FGF21. Recently, it has been extensively studied and has become a potential drug target for the treatment of metabolic diseases. As it is a protein-based hormone, FGF21 cannot be easily and quickly absorbed into the blood through oral administration. Moreover, it has a 0-2 h half-life in vivo, as shown in a previous study, thus its efficacy lasts for a short period of time when used to treat metabolic diseases, limiting its clinical applications. To avoid these limitations, we used Lactococcus lactis, a food-grade bacterium, as the host to express FGF21. It could be used successfully for the expression and long-term effect of FGF21 in vivo. Instead of antibiotic resistance genes, the LacF gene was used as a selection marker in the NZ3900/PNZ8149 expression system, which is safe and could reduce the antibiotic resistance crisis. In this study, we a constructed human FGF21 expressing L. lactis strain and administered it to Db/Db mice by gavage. Compared with the control group, the body weight of mice in the experimental group was significantly reduced, and the overall homeostasis was improved in mice treated with human FGF21. Moreover, the activity of brown adipose tissue was enhanced. These results revealed that oral administration of FGF21 through heterologous expression in L. lactis appears to be an effective approach for its clinical application.
Sigma-Aldrich
Nisin Ready Made Solution, (20,000-40,000 IU/mL in 0.02N HCl)