Merck
  • ATM-CHK2-Beclin 1 axis promotes autophagy to maintain ROS homeostasis under oxidative stress.
ATM-CHK2-Beclin 1 axis promotes autophagy to maintain ROS homeostasis under oxidative stress.
The EMBO journal (2020-03-19)
Qi-Qiang Guo, Shan-Shan Wang, Shan-Shan Zhang, Hong-De Xu, Xiao-Man Li, Yi Guan, Fei Yi, Ting-Ting Zhou, Bo Jiang, Ning Bai, Meng-Tao Ma, Zhuo Wang, Yan-Ling Feng, Wen-Dong Guo, Xuan Wu, Gui-Feng Zhao, Guang-Jian Fan, Sheng-Ping Zhang, Chuan-Gui Wang, Long-Yue Cao, Brian P O'Rourke, Shi-Hui Liu, Ping-Yuan Wang, Shuai Han, Xiao-Yu Song, Liu Cao
ABSTRACT
The homeostatic link between oxidative stress and autophagy plays an important role in cellular responses to a wide variety of physiological and pathological conditions. However, the regulatory pathway and outcomes remain incompletely understood. Here, we show that reactive oxygen species (ROS) function as signaling molecules that regulate autophagy through ataxia-telangiectasia mutated (ATM) and cell cycle checkpoint kinase 2 (CHK2), a DNA damage response (DDR) pathway activated during metabolic and hypoxic stress. We report that CHK2 binds to and phosphorylates Beclin 1 at Ser90/Ser93, thereby impairing Beclin 1-Bcl-2 autophagy-regulatory complex formation in a ROS-dependent fashion. We further demonstrate that CHK2-mediated autophagy has an unexpected role in reducing ROS levels via the removal of damaged mitochondria, which is required for cell survival under stress conditions. Finally, CHK2-/- mice display aggravated infarct phenotypes and reduced Beclin 1 p-Ser90/Ser93 in a cerebral stroke model, suggesting an in vivo role of CHK2-induced autophagy in cell survival. Taken together, these results indicate that the ROS-ATM-CHK2-Beclin 1-autophagy axis serves as a physiological adaptation pathway that protects cells exposed to pathological conditions from stress-induced tissue damage.
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Sigma-Aldrich
CHK2 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)
Sigma-Aldrich
Anti-Chk2 Antibody, clone 7, clone 7, Upstate®, from mouse