Merck
  • Cigarette smoke-induced lung inflammation in COPD mediated via CCR1/JAK/STAT /NF-κB pathway.
Cigarette smoke-induced lung inflammation in COPD mediated via CCR1/JAK/STAT /NF-κB pathway.
Aging (2020-05-29)
Kaishun Zhao, Ran Dong, Yanfang Yu, Chunlin Tu, Ying Li, YuJuan Cui, Lei Bao, Chunhua Ling
ABSTRACT
Inflammation is an important cause of chronic obstructive pulmonary disease (COPD) and its acute exacerbation. However, the critical role of C-C chemokine receptor (CCR)1 in progression of cigarette smoke-induced chronic inflammation remains unclear. We studied CCR1 expression using immunohistochemistry, immunofluorescence, and real-time polymerase chain reaction (RT-PCR) in COPD patients and controls. Cytokine levels in peripheral blood were measured by enzyme-linked immunosorbent assay (ELISA). In vitro, we investigated Janus kinase/signal transducers and activators of transcription (JAK/STAT)/nuclear factor-κB (NF-κB) signaling in cigarette smoke extract-induced or CCR1 deficiency/overexpressed mouse macrophage cell line MH-S by RT-PCR and western blot, and measured the cytokine levels in the supernatant with ELISA. We found that CCR1 expression was upregulated in COPD patients and there was a negative correlation between CCR1 mRNA levels and predicted % forced expiratory volume in 1 min. Inflammatory cytokine levels in the peripheral blood were higher in COPD patients than controls, and these were positively correlated with CCR1 levels. CCR1 was shown to play a critical role in regulating smoke-induced inflammation via JAK/STAT3/NF-κB signaling in vitro. CCR1 may play a critical role in airway inflammation in COPD. Additionally, understanding the molecular mechanism may help develop novel methods for the treatment of COPD.
Sigma-Aldrich
MISSION® esiRNA, targeting human LNPK
Sigma-Aldrich
MISSION® esiRNA, targeting human CCR1