Merck
  • A Photo-clickable ATP-Mimetic Reveals Nucleotide Interactors in the Membrane Proteome.
A Photo-clickable ATP-Mimetic Reveals Nucleotide Interactors in the Membrane Proteome.
Cell chemical biology (2020-06-11)
Mark Jelcic, Ke Wang, King Lam Hui, Xiao-Chuan Cai, Balázs Enyedi, Minkui Luo, Philipp Niethammer
ABSTRACT
ATP is an important energy metabolite and allosteric signal in health and disease. ATP-interacting proteins, such as P2 receptors, control inflammation, cell death, migration, and wound healing. However, identification of allosteric ATP sites remains challenging, and our current inventory of ATP-controlled pathways is likely incomplete. Here, we develop and verify mipATP as a minimally invasive photoaffinity probe for ATP-interacting proteins. Its N6 functionalization allows target enrichment by UV crosslinking and conjugation to reporter tags by "click" chemistry. The additions are compact, allowing mipATP to completely retain the calcium signaling responses of native ATP in vitro and in vivo. mipATP specifically enriched for known nucleotide binders in A549 cell lysates and membrane fractions. In addition, it retrieved unannotated ATP interactors, such as the FAS receptor, CD44, and various SLC transporters. Thus, mipATP is a promising tool to identify allosteric ATP sites in the proteome.
Sigma-Aldrich
Iodoacetamide-13C2, 2-d2, 99 atom % 13C, 98 atom % D
Sigma-Aldrich
Copper(II) sulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
Ethyl 3-aminobenzoate methanesulfonate, 98%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Guanosine 5′-triphosphate sodium salt hydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%
Sigma-Aldrich
Sodium molybdate, ≥98%
Sigma-Aldrich
Cytidine 5′-triphosphate disodium salt, ≥95%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt hydrate, Type IV, ≥93.0% (HPLC)
Sigma-Aldrich
Uridine 5′-triphosphate trisodium salt hydrate, from yeast, Type III, ≥96%
Sigma-Aldrich
L-Lysine monohydrate, BioReagent, suitable for cell culture, from non-animal source
Sigma-Aldrich
Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate, ≥93% (HPLC), powder
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
Adenosine 5′-diphosphate sodium salt, bacterial, ≥95% (HPLC)
Sigma-Aldrich
p-Toluenesulfonic acid monohydrate, ACS reagent, ≥98.5%
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, 99%
Sigma-Aldrich
Durcupan ACM, set for 1 L embedding mixture
Sigma-Aldrich
Sodium hydrosulfite, technical grade