Merck
  • Affibody-Conjugated RALA Polymers Delivering Oligomeric 5-Fluorodeoxyuridine for Targeted Therapy of HER2 Overexpressing Gastric Cancer.
Affibody-Conjugated RALA Polymers Delivering Oligomeric 5-Fluorodeoxyuridine for Targeted Therapy of HER2 Overexpressing Gastric Cancer.
Macromolecular bioscience (2020-06-20)
Fanghua Zhang, Jiwei Yin, Chao Zhang, Mengnan Han, Xuming Wang, Shuangqing Fu, Jie Du, Honglei Zhang, Wei Li
ABSTRACT
Affibody-conjugated RALA (affi-RA) are designed for delivering oligomeric 5-fluorodeoxyuridine (FUdR, metabolite of 5-FU) strand to raise the selectivity of 5-fluorouracil (5-FU), decrease its toxicity and improve its suboptimal therapeutic efficacy. The nanodrugs, FUdR@affi-RA, are spontaneously assembled by electrostatic interaction between positively charged affi-RA and negatively charged FUdR15 -strands (15 consecutive FUdR). FUdR@affi-RA exhibits excellent stability under simulated physiological conditions. Compared with free FUdR, FUdR@affi-RA shows excellent targeting and higher cytotoxicity in human epidermal growth factor receptor 2 (HER2) overexpressing gastric cancer N87 cells. Moreover, the anticancer mechanism studies reveal that FUdR@affi-RA enhances the expression and activity of apoptosis-associated proteins in the Bcl-2/Bax-caspase 8,9-caspase 3 apoptotic pathway induced by FUdR. This study indicates that the fusion vector, affi-RA, presents a promising delivery system platform for nucleoside analogue drugs and provides a new strategy for the development of therapeutics of cancer treatment.
Product Description
Supelco
Acetic acid – triethylamine solution 1:1, suitable for HPLC, 2M:2M aqueous solution, LiChropur
Pricing and availability is not currently available.