Merck
BRD4 jump-starts transcription after mitotic silencing.
Genome biology (2011-12-01)
Philipp Voigt, Danny Reinberg
ABSTRACT
The chromatin adapter BRD4 may be crucial for transmitting epigenetic information by acting as a histone acetylation-dependent gene bookmark and accelerating post-mitotic transcriptional reactivation.
Sigma-Aldrich
BRD4, BD1, BD2 (49-460) His tag human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
BRD4 (49-170) GST tag human, recombinant, expressed in E. coli, ≥82% (SDS-PAGE)
Sigma-Aldrich
BRD4 (342-460) GST tag human, recombinant, expressed in E. coli, ≥75% (SDS-PAGE)
Sigma-Aldrich
BRD4 (49-170) His tag human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Sigma-Aldrich
BRD4 (342-460) His tag human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)