Merck
  • RNF20 inhibits TFIIS-facilitated transcriptional elongation to suppress pro-oncogenic gene expression.
RNF20 inhibits TFIIS-facilitated transcriptional elongation to suppress pro-oncogenic gene expression.
Molecular cell (2011-05-21)
Efrat Shema, Jaehoon Kim, Robert G Roeder, Moshe Oren
ABSTRACT
hBRE1/RNF20 is the major E3 ubiquitin ligase for histone H2B. RNF20 depletion causes a global reduction of monoubiquitylated H2B (H2Bub) levels and augments the expression of growth-promoting, pro-oncogenic genes. Those genes reside preferentially in compact chromatin and are inefficiently transcribed under basal conditions. We now report that RNF20, presumably via H2Bub, selectively represses those genes by interfering with chromatin recruitment of TFIIS, a factor capable of relieving stalled RNA polymerase II. RNF20 inhibits the interaction between TFIIS and the PAF1 complex and hinders transcriptional elongation. TFIIS ablation selectively abolishes the upregulation of those genes upon RNF20 depletion and attenuates the cellular response to EGF. Consistent with its positive role in transcription of pro-oncogenic genes, TFIIS expression is elevated in various human tumors. Our findings provide a molecular mechanism for selective gene repression by RNF20 and position TFIIS as a key target of RNF20's tumor suppressor activity.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
RNF20 active human, recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Histone H2B Antibody, Upstate®, from rabbit