Merck
  • Silencing hepatic MCJ attenuates non-alcoholic fatty liver disease (NAFLD) by increasing mitochondrial fatty acid oxidation.
Silencing hepatic MCJ attenuates non-alcoholic fatty liver disease (NAFLD) by increasing mitochondrial fatty acid oxidation.
Nature communications (2020-07-06)
Lucía Barbier-Torres, Karen A Fortner, Paula Iruzubieta, Teresa C Delgado, Emily Giddings, Youdinghuan Chen, Devin Champagne, David Fernández-Ramos, Daniela Mestre, Beatriz Gomez-Santos, Marta Varela-Rey, Virginia Gutiérrez de Juan, Pablo Fernández-Tussy, Imanol Zubiete-Franco, Carmelo García-Monzón, Águeda González-Rodríguez, Dhaval Oza, Felipe Valença-Pereira, Qian Fang, Javier Crespo, Patricia Aspichueta, Frederic Tremblay, Brock C Christensen, Juan Anguita, María Luz Martínez-Chantar, Mercedes Rincón
ABSTRACT
Nonalcoholic fatty liver disease (NAFLD) is considered the next major health epidemic with an estimated 25% worldwide prevalence. No drugs have yet been approved and NAFLD remains a major unmet need. Here, we identify MCJ (Methylation-Controlled J protein) as a target for non-alcoholic steatohepatitis (NASH), an advanced phase of NAFLD. MCJ is an endogenous negative regulator of the respiratory chain Complex I that acts to restrain mitochondrial respiration. We show that therapeutic targeting of MCJ in the liver with nanoparticle- and GalNAc-formulated siRNA efficiently reduces liver lipid accumulation and fibrosis in multiple NASH mouse models. Decreasing MCJ expression enhances the capacity of hepatocytes to mediate β-oxidation of fatty acids and minimizes lipid accumulation, which results in reduced hepatocyte damage and fibrosis. Moreover, MCJ levels in the liver of NAFLD patients are elevated relative to healthy subjects. Thus, inhibition of MCJ emerges as an alternative approach to treat NAFLD.
Sigma-Aldrich
Eosin Y Solution, Aqueous
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Mayer′s Hematoxylin Solution
Sigma-Aldrich
Harris Hematoxylin Solution, Modified
Sigma-Aldrich
DPX Mountant for histology, slide mounting medium