- Up-regulation of CIR1/CROC1 expression upon cell immortalization and in tumor-derived human cell lines.
Up-regulation of CIR1/CROC1 expression upon cell immortalization and in tumor-derived human cell lines.
Acquisition of the immortal phenotype by tumor cells represents an essential and potentially rate-limiting step in tumorigenesis. To identify changes in gene expression that are associated with the early stages of cell immortalization, we compared genetically matched pairs of pre-immortal and immortal human cell clones by mRNA differential display. Two transcripts, denoted CIR1 and CIR2, were identified which were up-regulated in immortal cells. Sequence analysis revealed CIR1 to be identical to the recently cloned CROC1/UEV-1 gene, whereas CIR2 corresponds to an as yet uncharacterized 1.2 kb mRNA. A 5-6-fold elevation in CIR1/CROC1 expression and a 2-3-fold elevation in CIR2 expression were observed in SV40-transformed human embryonic kidney cells immediately following proliferative crisis, suggesting a potential role for these genes in immortalization. Expression of CIR1/CROC1 was found to be elevated also in a variety of immortal human tumor-derived cell lines, as compared to their normal tissue counterparts. These results are compatible with induction of CIR1/CROC1 being an early event in the acquisition of immortality and with a role for this gene in the immortal phenotype of tumor cells.