Merck
  • Wnt signaling activates MFSD2A to suppress vascular endothelial transcytosis and maintain blood-retinal barrier.
Wnt signaling activates MFSD2A to suppress vascular endothelial transcytosis and maintain blood-retinal barrier.
Science advances (2020-09-15)
Zhongxiao Wang, Chi-Hsiu Liu, Shuo Huang, Zhongjie Fu, Yohei Tomita, William R Britton, Steve S Cho, Chuck T Chen, Ye Sun, Jian-Xing Ma, Xi He, Jing Chen
ABSTRACT
Breakdown of the blood-retinal barrier (BRB) causes retinal edema and vision loss. We investigated the role of Wnt signaling in maintaining the BRB by limiting transcytosis. Mice lacking either the Wnt co-receptor low-density lipoprotein receptor-related protein 5 (Lrp5-/- ) or the Wnt ligand Norrin (Ndpy/- ) exhibit increased retinal vascular leakage and enhanced endothelial transcytosis. Wnt signaling directly controls the transcription of an endothelium-specific transcytosis inhibitor, major facilitator superfamily domain-containing protein 2a (MFSD2A), in a β-catenin-dependent manner. MFSD2A overexpression reverses Wnt deficiency-induced transcytosis in endothelial cells and in retinas. Moreover, Wnt signaling mediates MFSD2A-dependent vascular endothelium transcytosis through a caveolin-1 (CAV-1)-positive caveolae pathway. In addition, levels of omega-3 fatty acids are also decreased in Wnt signaling-deficient retinas, reflecting the basic function of MFSD2A as a lipid transporter. Our findings uncovered the Wnt/β-catenin/MFSD2A/CAV-1 axis as a key pathway governing endothelium transcytosis and inner BRB integrity.
Sigma-Aldrich
Anti-Rabbit IgG (H+L)-Peroxidase antibody produced in donkey, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
XAV939, ≥98% (HPLC)
Sigma-Aldrich
ANTI-FLAG® antibody, Rat monoclonal, clone 6F7, purified from hybridoma cell culture
Sigma-Aldrich
Peroxidase from horseradish, Type II, essentially salt-free, lyophilized powder, 150-250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for use with bacterial cell extracts, lyophilized powder