Merck
Advillin is a tuft cell marker in the mouse alimentary tract.
Journal of molecular histology (2020-07-04)
Anna-Lena Ruppert, Maryam Keshavarz, Sarah Winterberg, Johannes Oberwinkler, Wolfgang Kummer, Burkhard Schütz
ABSTRACT
Tuft cells are a rare population of chemosensory cells at the mucosal surface epithelia of hollow organs. Their name-giving morphological feature is an apical tuft of stiff microvilli. Accordingly, the actin-binding protein, villin, was identified as one of the first tuft cell markers in immunohistochemical analysis. Unfortunately, villin expression is not restricted to tuft cells, but is also prominent e.g. in enterocytes, which limits the use of this gene as a marker and as an experimental tool to genetically target tuft cells. Here, we report that the villin-related protein, advillin, is a specific tuft cell marker in the gastro-intestinal and biliary tract epithelia. In situ hybridization and immunohistochemistry revealed that advillin expression, unlike villin, was restricted to solitary cholinergic tuft cells in the mucosal linings of the small and large intestine, and in the gall bladder. In the glandular stomach, villin and advillin mRNA were present in all epithelial cells, while detectable protein levels were confined to solitary tuft cells. Advillin expression was no longer detectable in the mucosa of the intestinal and biliary tract from Pou2f3 deficient mice that lack tuft cells. Finally, crossing Avil-Cre transgenic mice with a double-fluorescent reporter mouse line resulted in specific targeting of gastro-intestinal and biliary tuft cells. Our analysis introduces advillin as a selective marker and tool in histological and functional analysis of the alimentary tract tuft cell system.
Sigma-Aldrich
Triethanolamine, reagent grade, 98%
Sigma-Aldrich
Acetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Dextran sulfate sodium salt from Leuconostoc spp., Mr ~500,000
Sigma-Aldrich
Hematoxylin cryst. (C.I. 75290), for microscopy
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Denhardt′s Solution, for Northern and Southern blotting, lyophilized powder
Sigma-Aldrich
Methyl Green, zinc chloride salt, <0.5% crystal violet, Dye content 80 %
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%