Merck
  • BAFF blockade prevents anti-drug antibody formation in a mouse model of Pompe disease.
BAFF blockade prevents anti-drug antibody formation in a mouse model of Pompe disease.
Clinical immunology (Orlando, Fla.) (2015-04-07)
Phillip A Doerfler, Sushrusha Nayak, Roland W Herzog, Laurence Morel, Barry J Byrne
ABSTRACT
Antibodies formed against the therapeutic protein are a life-threatening complication that arises during enzyme replacement therapy for Pompe disease (acid α-glucosidase deficiency; GAA). To provide an effective alternative to current practices, we investigated the capacity of anti-B-cell activating factor (BAFF) as a novel drug candidate to prevent antibody formation in a Pompe disease mouse model. A BAFF-neutralizing antibody was administered prophylactically and with maintenance doses in association with enzyme replacement therapy using recombinant human GAA in Gaa(-/-) mice. BAFF blockade delayed antibody production and increased GAA activity within tissues with protection from anaphylaxis. Anti-BAFF also resolved antibody formation during an immune response and precluded the maturation of antibody secreting cells from entering the bone marrow compartment. This treatment modality may therefore be a viable alternative for the clinical management of antibody formation for Pompe disease and has potential use against antibody formation in other protein replacement therapies.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
IgG1, Kappa from murine myeloma, clone MOPC 21, purified immunoglobulin, buffered aqueous solution