Merck
  • Radiobiologic effects of GS-nitroxide (JP4-039) on the hematopoietic syndrome.
Radiobiologic effects of GS-nitroxide (JP4-039) on the hematopoietic syndrome.
In vivo (Athens, Greece) (2011-05-18)
Julie P Goff, Michael W Epperly, Tracy Dixon, Hong Wang, Darcy Franicola, Donna Shields, Peter Wipf, Song Li, Xiang Gao, Joel S Greenberger
ABSTRACT
Total-body irradiation (TBI) doses in the range of 2-8 Gy are associated with a drop in peripheral blood counts, decreased bone marrow cellularity, and hematopoietic syndrome. Radiation mitigators must be safe for individuals likely to recover spontaneously. Female C57BL/6HNsd mice exposed to 9.0 and 9.15 Gy TBI, received intraperitoneal (10 mg/kg) JP4-039, a novel radiation mitigator, 24 hours after irradiation and were followed for hematopoietic recovery. Irradiated mice showed reduced peripheral blood lymphocytes and neutrophils and bone marrow cellularity at day 5. Serum electrolytes, liver and renal function tests showed no deleterious effect of JP4-039-after irradiation, and no reduction in survival compared to irradiated controls. Marrow recovery measured as cellularity, and hematopoietic colony-forming cells including primitive granulocyte-erythroid-megakaryocyte-monocytes (GEMM), reached pre-irradiation levels by day 30 in JP4-039 treated groups. Mice receiving single or multiple administrations of JP4-039 showed an early return of CFU-GEMM. JP4-039 (GS-Nitroxide) is a safe radiation mitigator in mice warranting studies in larger animals and potentially a Phase I Clinical Trial.
Sigma-Aldrich
JP4-039, ≥98% (HPLC)